About this item
BRANDplates® cellGrade™ premium plates are sterile plates treated for the cultivation of fastidious adherent cell lines
- poly-D-lysine-like surface for direct attachment of fastidious cell lines
- ANSI-SBS formatted
- Sterility assurance level of 10-6
All BRANDplates® are manufactured to ANSI/SLAS standards 1 to 4-2004 to ensure compatibility for all processes, especially when automation is involved. The 96-Well plates have an orange embossed alphanumeric code for quick and easy well identification.
The plate features a poly-D-lysine-like surface so cells can directly attach and spread out with optimal adhesion. Suitable for reduced serum or serum-free cultivation. Plates offer an excellent alternative to biologically coated surfaces. Plates are made of virgin polystyrene. Sterilized by beta irradiation according to Ph. Eur. and USP 29, to a Sterility Assurace Level of 10-6.