![WHEATON.jpg](https://occapi.avantorsciences.com/medias/WHEATON.jpg?context=bWFzdGVyfGltYWdlc3wxNTM5MTB8aW1hZ2UvanBlZ3xhR1V3TDJoaFpTODVORGd6TWpBMU5UUXhPVEU0TDFkSVJVRlVUMDR1YW5Cbnw3MzRiODc3ODA2NDIwYTZmYmY5YzQ2OWI1ZTFkZTBiMTUwMjVhN2E2MjBiYWMyNjgyODVlNDg3NTJkNTllZGM5)
![DWK_logo_40h.png](https://occapi.avantorsciences.com/medias/DWK-logo-40h.png?context=bWFzdGVyfGltYWdlc3wyNTIzfGltYWdlL3BuZ3xhR1V5TDJoaFlpODVORGd6TWpBMU5qQTNORFUwTDBSWFMxOXNiMmR2WHpRd2FDNXdibWN8YmQyM2M4ZDk3MDFiZTdlMDA4OTBlOTllOGU4NjAzZjRmZWNkYzRiZDgzMGQyNWMzNjk1NDhkYjhlMzZlYzViMQ)
Bottles for every lab
No matter the application, WHEATON has the right bottle for you
Product selection
WHEATON™ Media Bottles are a mainstay in laboratories across the world. All WHEATON Media Bottles with caps feature sloping shoulders to facilitate pouring and attached pour rings to eliminate dripping.
WHEATON Serum Bottles are offered in sizes that range from 2mL to 500mL. They are made from borosilicate glass that conforms to USP Type I requirements.
Leak-resistant, HDPE, LDPE, PP and much more.
As one of the world‘s top borosilicate glass, Duran® glass has proven itself excptional in both laboratories as well as in industrial applications.