Life Science
Use the recently updated Minimate™ EVO TFF System and Capsules to streamline laboratory-scale concentration, desalting, and buffer exchange processes in a simple to use plug-n-play benchtop configuration to process samples upto 1 L or more down to as little as 5 ml.
For high throughput applications, Pall’s AcroPrep™ portfolio offers 24, 96, and 384-well high-performance filter plates for rapid sample processing. A comprehensive selection of membranes, sample well volumes, and tip configurations allows researchers to select the best filter plate that fits their specific application needs.
Introducing the NAB Nanosep®, the first Universal Nucleic Acid Binding spin filter which can process most workflows in this one device. The NAB Nanosep includes a unique dual layer membrane enabling efficient and high recovery for downstream analysis.
- Enables recovery from 10,000 down to 50 base pairs
- Analyzes the widest range of nucleic acid fragments
- For Total RNA, pDNA, gDNA applications
- Processes up to 500uL of sample
- Generates enough sample to allow for multiple downstream testing
- Supported by a wide range of protocols for mammalian, bacteria, yeast, and plant starting materials (available upon request)
The NEW Acro™ 500 Capsule has been specifically designed for efficient, cost-effective filtration and high throughput of liquid volumes, remove particulate contamination, clarify solutions, and sterilize liquid solutions prior to further processing. The Acro™ 500 is uniquely housed in a clear polycarbonate housing; where easy observation is now possible to ensure peak performance by maximizing the entire effective filtration area (EFA) ideal for high turbidity, high bioburden solutions.
Product selection
Increase recovery, reduce processing time, reduced aggregation and precipitation while concentrating and/or buffer exchange with your nucleic acids and proteins.
A wide variety of plates to fit your high throughput needs for genomic and proteomic sample preparation
Easy-to-use system can process sample volumes up to 1L or more and efficiently concentrate samples to as little as 5 mL
Pall Laboratory offers a wide variety of products for your sterile filtration needs for filtration of media, buffers, supplements, samples, or media optimization projects.
Pall Laboratory offers the broadest variety of vent filters for liquid and gaseous laboratory applications
Pall offers a wide variety of membranes that provide high affinity for biomolecules and low background with many detection techniques.
For sterilization or clarification of laboratory fluids. Ideal for cell culture and serum-containing media.