![UV5-Excellence-955x590.jpg](https://occapi.avantorsciences.com/medias/UV5-Excellence-955x590.jpg?context=bWFzdGVyfGltYWdlc3wxMjg3MTB8aW1hZ2UvanBlZ3xhRGc0TDJobU15ODVORGd3TlRNME1qWXhOemt3TDFWV05TMUZlR05sYkd4bGJtTmxMVGsxTlhnMU9UQXVhbkJufGQ2NDYxNzZjNTBlYjkzMDI1MWJjNjIzODFjYzc5NzY1YzgyNGQzZWVjMjU2NGQwZGM3NTY1YjRjMGZkNDYyY2U)
![mettler_toledo_wordmark_150w.jpg](https://occapi.avantorsciences.com/medias/mettler-toledo-wordmark-150w.jpg?context=bWFzdGVyfGltYWdlc3wyMzk5fGltYWdlL2pwZWd8YURZeEwyZ3pNUzg1TkRnd05UTXlNREF3TnprNEwyMWxkSFJzWlhKZmRHOXNaV1J2WDNkdmNtUnRZWEpyWHpFMU1IY3VhbkJufDdlYjhhYTZhYWE1Y2Q4YjRjOTY1YjRlODc0ZjIzYjkxMGQ3MDFmMzE5MzZhOGYzOWMwZWZjZjQ3YmM3OGJiYTk)
UV/Vis Spectrophotometers
UV/VIS Excellence spectrophotometers combine outstanding optical performance based on FastTrack™ technology with the intuitive One Click™ user interface. These models include a state-of-the-art, long-life Xenon lamp for stable, repeatable and sustainable measurements, and are always ready for measurement, no warm-up time needed. These units have a compact design without compromising performance.
![1CPCTA-260x190.jpg](https://occapi.avantorsciences.com/medias/1CPCTA-260x190.jpg?context=bWFzdGVyfGltYWdlc3w0NjIyOHxpbWFnZS9qcGVnfGFETmpMMmhpWVM4NU5EZ3dOVE16T1RNME1URXdMekZEVUVOVVFTMHlOakI0TVRrd0xtcHdad3xiZGJlNDM0ODE2MmIwN2QxODAxOTI0Nzk2YmQ3NGMzOGIwYjA3NmZkNDdhZmIyNDAyZjQxZTM4OGFjNjJiODZj)
Current Promotions
- Receive up to a $400 cash-back rebate with your next METTLER TOLEDO Balance, pH Meter, or Analytical Instrument purchase.
- Trade in your old balance and receive a 15% cash back rebate with your new Balance purchase.
Product selection
Unique design with excellent optical performance.
A wide range of accessories turns your UV/Vis into a dedicated system to solve your application needs.
Streamline your UV/Vis workflow with LabX® Software.
Maximize performance with professional installation.