![lucigen_routine_pcr_feature.jpg](https://occapi.avantorsciences.com/medias/lucigen-routine-pcr-feature.jpg?context=bWFzdGVyfGltYWdlc3wzMDM0N3xpbWFnZS9qcGVnfGFEYzNMMmhqT0M4NU5qSXhPVGN5TnpRMk1qY3dMMngxWTJsblpXNWZjbTkxZEdsdVpWOXdZM0pmWm1WaGRIVnlaUzVxY0djfDFlMDdlNTI2NjJjMzM5OTJhN2FiYmQ0MGQwOTAxMWVkN2IwN2E5MDkwMjY4MWVhM2I0OGRmM2RjNDU3MzIxMGE)
![lucigen_lcg_biosearch_logos_40h.png](https://occapi.avantorsciences.com/medias/lucigen-lcg-biosearch-logos-40h.png?context=bWFzdGVyfHJvb3R8NjkzN3xpbWFnZS9wbmd8YUdWaEwyaGtNeTg1TkRjNU5UZzBOalExTVRVd0wyeDFZMmxuWlc1ZmJHTm5YMkpwYjNObFlYSmphRjlzYjJkdmMxODBNR2d1Y0c1bnwwMmMyNDgwM2U3YmYxZjNkOGU1YWJmZTU5NzVhMDI0ZDIwYThiMDU1MmY3MWQwZjQyODJlZjQ4ZjZjMDM4YTRm)
Routine PCR & Amplification Competent Cells, PCR & Amplification Enzymes
Competent Cells, PCR & Amplification Enzymes
EconoTaq® PLUS GREEN 2X Master Mix
- The easiest solution for PCR. Ready-to-use 2X Taq DNA Polymerase master mix formulated with a proprietary PCR enhancer for optimal performance, dNTPs, and two tracking dyes for easy gel loading and monitoring.
EconoTaq® PLUS 2X Master Mix
- Same great performance as EconoTaq PLUS GREEN, but does not contain tracking dyes, avoiding interference with fluorescence detection.
EconoTaq® DNA Polymerase
- High quality Taq DNA polymerase at a low price.
CloneID™ 1X Colony PCR Mix
- Convenient and powerful 1X master mix for direct Colony PCR.
Bst DNA Polymerase, Exonuclease Minus
- Isothermal amplification and strand displacement for LAMP and NextGen sequencing.