![Leica-THUNDER955-590.gif](https://occapi.avantorsciences.com/medias/Leica-THUNDER955-590.gif?context=bWFzdGVyfHJvb3R8MjI2NDI2fGltYWdlL2dpZnxhR1F5TDJneVl5ODVORGM1TmpBNU1EVTNNekV3TDB4bGFXTmhMVlJJVlU1RVJWSTVOVFV0TlRrd0xtZHBaZ3xkMTQ5YjRjZjg4MDZhNThkMjY3MDJhMDQ0OTEyMGNkOTdjY2IwNGExNDgyMmI5ZjEwM2UxMjE4ODMzZGM5ZGE3)
![leica_microsystems_logo_4.jpg](https://occapi.avantorsciences.com/medias/leica-microsystems-logo-4.jpg?context=bWFzdGVyfHJvb3R8MjEzM3xpbWFnZS9qcGVnfGFEWTNMMmd5WXk4NU5EYzVOakE1TURrd01EYzRMMnhsYVdOaFgyMXBZM0p2YzNsemRHVnRjMTlzYjJkdlh6UXVhbkJufGEwNjQ4MzlmMDcwNDQzNTJhN2ExZjMwZDAzMzRiOWEwMzU5NDgwNDNjYTM4NTZmMGUwYjBhZDVlNDZhMDJmODQ)
Leica THUNDER Imagers
To answer important scientific questions, they enable you to obtain a clear view of details, even deep within an intact sample, in real time without out-of-focus blur. Sharp imaging of 3D specimens is now as easy as working with your favorite camera-based fluorescence microscope. THUNDER Imagers with Computational Clearing define a new class of instruments for high-speed, high-quality imaging of thick, 3-dimensional specimens.
Product selection
THUNDER Imagers provide you with a solution for advanced 3D cell culture assays, whether you want to study stem cells, spheroids, or organoids.
The THUNDER Imager Model Organism allows fast and easy 3D exploration of whole organisms for developmental or molecular biology research.
The THUNDER Imager Tissue allows real-time fluorescence imaging of 3D tissue sections typically used in neuroscience and histology research. Acquire rich, detailed images of thick tissues free of haze from out-of-focus blur.