![microscopy-sample-feature.jpg](https://occapi.avantorsciences.com/medias/microscopy-sample-feature.jpg?context=bWFzdGVyfGltYWdlc3wxMjg4OTF8aW1hZ2UvanBlZ3xhR1kyTDJnNFlpODVOVGszTVRFeU1qSXhOekkyTDIxcFkzSnZjMk52Y0hrdGMyRnRjR3hsTFdabFlYUjFjbVV1YW5CbnxkYzhhMDkyNjFkMDg3MDkzZmFlYmRlMWY1N2I3MjBkY2JhYWI1ZDU1YTYyMzlmMDU2YWJmYmE4ZjI0ZjU3MDhi)
![leica_microsystems_logo_40h.png](https://occapi.avantorsciences.com/medias/leica-microsystems-logo-40h.png?context=bWFzdGVyfHJvb3R8MjEzM3xpbWFnZS9wbmd8YURCbUwyZ3laaTg1TkRjNU5qQTFNVEkxTVRVd0wyeGxhV05oWDIxcFkzSnZjM2x6ZEdWdGMxOXNiMmR2WHpRd2FDNXdibWN8YWU0ZWJlM2I5N2ExNjljOTU5NTAzNzFmMmZmN2MwZTA3ODhhNDAwOTQ2MDg3OTg5M2YxYzA2NTU3NDBiODA3OA)
Leica Electron Microscopy Sample Preparation
In the field of electron microscopy, perfect sample preparation is a prerequisite and crucial step. Leica Microsystems offers a comprehensive product portfolio for preparation of biological, medical and industrial samples...
Ensure the ultrastructures of your tissue samples can be precisely compared every time by preparing them with the EM TP tissue processor.
With the Electron Microscopy Tissue Processer (EM TP) you can be confident that tissue differences between samples observed with your light microscope (LM) or electron microscope (EM) are not caused by inconsistent manual preparation. The EM TP limits manual handling by at least 75% due to programmable, automatic processing of multiple samples.
Product selection
With the Leica EM TP, all processing steps of your experiment can be programmed with ease and are then executed by the system automatically. Trust in the comparability of your results and save valuable time with each run.