Orders LinkContactUsLinkComponent
Halo_environment955-590.png
HALO® HPLC Columns for Environmental Separations
HALO® ENVIROCLASS is the newest family of products in Advanced Materials Technology’s product line.
From performance-designed application-specific phases to separate per-and polyfluorinated alkyl substances (PFAS) and polycyclic aromatic hydrocarbons (PAH), HALO® ENVIROCLASS represents more than method assured products.
It offers a suite of solutions for other persistent, high environmental-impact contamination agents such as pesticides, mycotoxins, herbicides, and more all based upon HALO’s innovative and trusted technology all with the understanding of the unique needs of the environmental laboratory
Environmental Columns Literature
HALO HPLC Columns for Environmental Separations | Avantor
HALO® PFAS Analysis Solutions
Designed for separation of novel and legacy short chain and long chain PFAS compounds containing branched and linear isomers, along with EPA methodology requirements in mind, HALO® PFAS offers a holistic solution.
A PFAS specific delay column is optimized to prevent background PFAS contamination from interfering with the sample results. HALO® PFAS solution delivers excellent selectivity, peak shape and necessary retention to perform fast, high-resolution separations
Features
  • Application-assured through method qualified lot analysis
  • Endcapped alkyl phases for high sensitivity LCMS analysis
  • Optimal 2.7 µm Fused-Core® particle for rugged, reliable performance delivering high efficiency, low back pressure separations
HALO® PAH Analysis Solution:
The HALO® PAH column is an un-endcapped trifunctional C18 bonded phase with a proprietary manufacturing process designed on proven Fused-Core® technology to provide a fast, efficient separation of PAH compounds.
The HALO® PAH column delivers a method-specific, robust, high efficiency separation of 16 standard PAH compounds with a resolution value of at least 1.5 in under 5 minutes for EPA 8310 and EPA 610
Features
  • Application-assured through method qualified lot analysis
  • Optimal 2.7 µm Fused-Core® particle for rugged, reliable performance delivering high efficiency, low back pressure separations
  • Well suited for UV, Fluorescence and MS detection