![HaloColumns_Header.jpg](https://occapi.avantorsciences.com/medias/HaloColumns-Header.jpg?context=bWFzdGVyfGltYWdlc3wxMDA4NjR8aW1hZ2UvanBlZ3xhREkyTDJoaU5pODVOVEl4TWpnME9UQTNNRE00TDBoaGJHOURiMngxYlc1elgwaGxZV1JsY2k1cWNHY3w0NDIwZDRiMjY2YmQxNzQwNThmN2VmYmMwOTUyMzQxNDUxZmNhMThlZTI1MDczY2MzYmJjZTRmNTUzMTU1MmI3)
HALO® 1.5 MM ID HPLC Columns
The New HALO® 1.5 - Defining a New Dimension in Chromatography
The adoption of UHPLC instrumentation pushed LC separations to a new level of speed and resolution.
In the years since, chromatographers continue to push separation limits in response to increasing challenges. Now in 2022 AMT breaks new ground again with a 1.5 mm internal diameter HALO® column to push the boundaries of adopted UHPLC systems. Founded on all of the benefits of Fused-Core® particles, the HALO® 1.5 delivers increased sensitivity and reduced solvent consumption, allowing scientists to experience the benefits of capillary columns without the pains of specialized microflow systems.
Delivering More Performance
- More Sensitivity from conventional UHPLC Systems
- Higher Ionization Efficiencies from LCMS systems
- Reduce Solvent Consumption compared to 2.1 mm ID columns
- Easy to Implement microflow solution
1.5 MM ID Columns
- HALO 160 A ES-C18 2.7 UM 1.5 X 50 MM
- HALO 160 A ES-C18 2.7 UM 1.5 X 100 UM
- HALO 160 A ES-C18 2.7UM 1.5 X 150 MM
- HALO 1000 A C4 2.7 UM 1.5 X 50 MM
- HALO 1000 A DIPHENYL 2.7 UM 1.5 X 50 MM
- HALO 1000 A C4 2.7 UM 1.5 X 100 MM
- HALO 1000 A C4 2.7 UM 1.5 X 150 MM
- HALO 1000 A DIPHENYL 2.7 UM 1.5 X 150 MM
- HALO 90 A C18 2.7 UM 1.5 X 50 MM
- HALO 90 A C18 2.7 UM 1.5 X 100 MM
- HALO 90 A C18 2.7 UM 1.5 X 150 MM