Orders LinkContactUsLinkComponent
tissue-culture955x590.jpg
Greiner Bio-One CELLSTAR® Tissue Culture
Greiner Bio-One CELLSTAR® Tissue Culture | Avantor
Greiner Bio-One CELLSTAR® products offer a wide spectrum of cell culture flasks, dishes and plates. Their functional design, particular material properties and special coatings enable these vessels to be used not only in standard cell culture but also in cancer research, viral diagnostics, genetic engineering and vaccine production.
In response to the increasing automation of cell culture procedures, Greiner Bio-One developed the AutoFlask, a cell culture flask for automated systems. With the ThinCertPlate and appropriate cell culture inserts, Greiner Bio-One has also created an innovative platform for compatibility tests using skin models in the chemical and cosmetic industries.
The Cell Culture product sector was recently expanded with the innovative Advanced TC polymer modification. These cell culture products have a special surface that enables more effective cultivation of fastidious cells and facilitates complex testing systems
For high-resolution microscopy and live cell analysis, Greiner Bio-One offers the new CELLview dish, a polystyrene cell culture dish with an embedded autofluorescence-free glass bottom. The unique subdivided version of the dish is ideally suited for multiplex analyses, for example, and is also available with the Advanced TC surface.
Product selection
greiner_cell_culture_roller_bottles_90.jpg
CELLMASTER™ roller bottles are made from polystyrene (PS) or polyethylene terephthalate (PET). All roller bottles are sterilized by irradiation.
greiner_cellcoat_protein_90.jpg
CELLCOAT® cell culture vessels coated with proteins of the extracellular matrix or synthetic proteins. Improved adhesion and proliferation of primary cells and various cell lines
greiner_cellstar_cell_repellant_90.jpg
CELLSTAR® cell culture vessels with a cell-repellent surface prevent cell attachment in suspension cultures of semi adherent and adherent cell lines.
greiner_cell_culture_microscopy_90.jpg
CELLview cell culture dish combined 35 mm disposable plastic cell culture dish with the optical quality of glass, providing superior high-resolution microscopic images of in-vitro cultivated cultures.
greiner_cell_scrapers_90.jpg
Pivoted scraper blade guarantees uniform contact with the growth surface, minimal mechanical strain and efficient cell harvest, even from poorly accessible vessels. 
greiner_easystrainer_100.jpg
EASYstrainer Cell Sieves, Greiner Bio-One
EASYstrainer™ cell strainers are filters for Cell suspensions and primary cell isolates. Sterile. Fits all standard 50ml tubes. Venting slot for fast filtration
greiner_thincert_90.jpg
ThinCert™ Cell Culture Inserts
ThinCert cell culture inserts suitable for transport, secretion and diffusion studies, migrational experiments, cytotoxicity testing, co-cultures, as primary cell culture
greiner_leucosep_tubes_90.jpg
Leucosep™ Centrifuge Tubes, Greiner Bio-One
Leucosep developed for optimal separation of lymphocytes and peripheral mononuclear cells from human whole blood and bone marrow. Key feature is the porous barrier incorporated in the centrifuge tube.
greiner_cryos_90.jpg
CYROs
The portfolio of Cryo.s tubes includes cryogenic tubes with different volumes, different base forms as well as several cap colors.
greiner_serologicals_100.jpg
A wide range of different serological pipettes. The volume capacity of the pipettes is increased by a negative graduation. The pipettes are provided with color codes according to international standards
3D_Cell_Culturing_90x90.jpg
The core technology is the magnetization of cells with NanoShuttle-PL. The cells can be aggregated with magnetic forces, either by levitation or printing, to form structurally and biologically representative 3D models in vitro.
greiner_surfaces_100.jpg
CELLSTAR® offers a wide spectrum of cell culture flasks, dishes and plates. Used in standard cell culture and in cancer research, viral diagnostics, genetic engineering and vaccine production
greiner_advanced_tc_vessels_90.jpg
Advanced TC Cell Culture Vessels for the propagation of fastidious cells like primary or sensitive cells as well as cells cultivated under restricted growth conditions.