Magnetic 3D Cell Culturing
![cell-culture-3D-955x590.jpg](https://occapi.avantorsciences.com/medias/cell-culture-3D-955x590.jpg?context=bWFzdGVyfGltYWdlc3w1Njk1OXxpbWFnZS9qcGVnfGFHVTBMMmd3TUM4NU5qQTJNRFUyTkRFeE1UWTJMMk5sYkd3dFkzVnNkSFZ5WlMwelJDMDVOVFY0TlRrd0xtcHdad3w2NzMxNTk3Y2IyYzY0YTQxMzI0NGMzMWE3OTBlNTM0YmUyZmZlMTdiODEwZWQ3NzMwZmM0NTk5NWUxMzEzZjRi)
Magnetic 3D Cell Culturing 3D in a fast and easy 2D workflow
The core technology is the magnetization of cells with NanoShuttle™-PL. The cells can be aggregated with magnetic forces, either by levitation or printing, to form structurally and biologically representative 3D models in vitro.
- The advantages of magnetic cell culture include
- Mimicking native tissue environment
- Rapid 3D model formation within hours
- No specialized equipment, media, or artificial substrate
- Easy to handle / no sample loss
- Allows co-culture
![Cell-Repellent-Surface-955x590.jpg](https://occapi.avantorsciences.com/medias/Cell-Repellent-Surface-955x590.jpg?context=bWFzdGVyfGltYWdlc3w1NTYzM3xpbWFnZS9qcGVnfGFETTJMMmd3TUM4NU5qQTJNRFUyTkRjMk56QXlMME5sYkd3dFVtVndaV3hzWlc1MExWTjFjbVpoWTJVdE9UVTFlRFU1TUM1cWNHY3xkYzczOWZiNDZkN2YyMzdiNGE5ZjAyZWVkNzM1YmE5NzEzY2RlN2UxMjIzYzQzMTJjMWU1OTI1YzUxNzI0NjQ5)
CELLSTAR® Cell Culture Vessels with Cell-Repellent Surface
In contrast to standard tissue culture surfaces which are optimized to enhance conditions for cell attachment, the cell-repellent surface has been developed to effectively prevent cell attachment. CELLSTAR® cell culture vessels with a cell-repellent surface reliably prevent cell attachment in suspension cultures of semi adherent and adherent cell lines where standard hydrophobic surfaces generally used for suspension culture are insufficient.
![cell-culture-PDF-front-260x190.jpg](https://occapi.avantorsciences.com/medias/cell-culture-PDF-front-260x190.jpg?context=bWFzdGVyfGltYWdlc3w5MTM0fGltYWdlL2pwZWd8YUdKa0wyaG1OUzg1TmpFNU9URTRPVGM1TVRBeUwyTmxiR3d0WTNWc2RIVnlaUzFRUkVZdFpuSnZiblF0TWpZd2VERTVNQzVxY0djfDA0MTg1ZGFhZTMzNjkzMjhjOThhZTA1MmZjNzcwMmZiODFiMGE3NTZiMzhjOTM2ZjNjNjkwMzQ3MmY0YzJiODA)
3D Cell Culture Literature
Magnetic 3D Cell Culture Products
VWR PN | GBO PN | Product Descriptions |
76324-742 | 657840 | 6-Well Bio-Assembler Kit |
76324-760 | 662840 | 24-Well Bio-Assembler™ Kit |
76324-014 | 655840 | 96-Well Bioprinting Kit, Clear Plates |
76324-016 | 655841 | 96-Well Bioprinting Kit, Black µClear Plates |
76324-018 | 655846 | 96-Well BiO Assay™ Kit |
76324-020 | 655849 | 96-Well Bio Assay Kit™ & Imaging System |
76324-768 | 781840 | 384-Well Bioprinting Kit, Clear Plates |
76324-770 | 781841 | 384-Well Bioprinting Kit, Black µClear Plates |
76324-772 | 781846 | 384-Well BiO Assay™ Kit |
76324-282 | 781849 | 384-Well BiO Assay™ Kit & Imaging System |
76337-248 | 781850 | 384-Well Ring Drive |
76337-250 | 789830 | 1536 well Spheroid Drive |
76337-252 | 789837 | 1536 WELL HOLDING DRIVE |
76324-744 | 657841 | NanoShuttle™-PL Refill, 600 µL Vial (1) |
76324-746 | 657843 | NanoShuttle™-PL Refill 3 Pack, 600 µL Vials (3) |
76324-748 | 657846 | NanoShuttle™-PL Refill 6 Pack, 600 µL Vials (6) |
76324-750 | 657847 | NanoShuttle™-PL Refill 6 Pack with FREE iPod™ (Only with purchase of BIO Assay™ 655849 or 781849) |
76324-754 | 657852 | NanoShuttle™-PL Refill 12 pack, 600 µl Vials |
76324-752 | 657850 | MagPen™ 3 Pack |
76324-024 | 657860 | Imaging Kit |
CELLSTAR® Cell Culture Vessels with Cell-Repellent Surface
VWR PN | GBO PN | Product Descriptions |
10014-312 | 627979 | CELLSTAR 35mm dish, cell-repellent surface, clear, sterile, with lid |
10014-314 | 628979 | CELLSTAR® Cell-Repellent Surface, 60x15mm, sterile, with lid |
30618-024 | 664970 | Cellstar 100mm dish, cell-repellent surface, clear, sterile, with lid |
75838-906 | 690980 | CELL CULTURE FLASK, 50 ML, 25 CM², PS, CLEAR,CELLSTAR®, CELL-REPELLENT SURFACE, WHITE STANDARD SCREW CAP, STERILE, 10 PCS./BAG |
10836-539 | 690985 | T25 CELLSTAR® Cell-Repellent Surface, sterile, filter cap |
75838-908 | 658980 | CELL CULTURE FLASK, 250 ML, 75 CM², PS,CLEAR, CELLSTAR®, CELL-REPELLENT SURFACE, WHITE STANDARD SCREW CAP, STERILE, 5 PCS./BAG |
10836-541 | 658985 | T75 CELLSTAR® Cell-Repellent Surface, sterile, filter cap |
75845-114 | 660980 | CELL CULTURE FLASK, 550 ML 175 CM², PS, Clear, CELLSTAR®, Cell Repellent Surface, White Standard Screw Cap |
75800-006 | 660985 | CELL CULTURE FLASK, 550 ML, 175 CM², PS, Clear, CELLSTAR®, Cell Repellent Surface, White Filter Screw Cap, Sterile |
75838-910 | 661980 | CELL CULTURE FLASK, 650 ML, 175 CM², PS,HIGH FLASK DESIGN, CLEAR, CELLSTAR®, CELL-REPELLENT SURFACE, WHITE STANDARD SCREW CAP,STERILE, 4 PCS./BAG |
30618-022 | 657970 | Cellstar 6 well plate, cell-repellent surface, clear, sterile, with lid |
10014-320 | 662970 | 24 Well CELLSTAR® Cell-Repellent Surface, sterile, with lid |
10014-322 | 677970 | 48 Well CELLSTAR® Cell-Repellent Surface, sterile, with lid |
30618-028 | 655970 | 96W Cell-Repellent Plate, PS, Sterile, Flat Bottom, Chimney Style, Clear, w/Lid |
10014-318 | 655976 | 96W Cell-Repellent Plate, PS, Sterile, Flat Bottom, Black µClear, w/Lid |
10859-852 | 655976-SIN | 96W Cell-Repellent Plate, PS, Sterile, Flat Bottom, Black µClear, w/Lid |
30618-026 | 650970 | 96W Cell-Repellent Plate, PS, Sterile, Round (U) Bottom, Clear, w/Lid |
76199-200 | 650979 | 96W Cell-Repellent Plate, PS, Sterile, Round (U) Bottom, Clear, w/Lid |
10014-316 | 651970 | 96W Cell Repellent Plate, PS, Sterile, Conical (V) Bottom, Clear, w/Lid |
10014-324 | 781970 | 384W Cell-Repellent Plate, PS, Sterile, Flat Bottom, Clear, w/Lid |
76371-716 | 781974 | 384W Cell-Repellent Plate, PS, Sterile, Flat Bottom, White µClear, w/Lid |
10014-326 | 781976 | 384W Cell-Repellent Plate, PS, Sterile, Flat Bottom, Black µClear, w/Lid |
10859-850 | 781976-SIN | 384W Cell-Repellent Plate, PS, Sterile, Flat Bottom, Black µClear, w/Lid |
76199-198 | 787979 | 384 Well Cell-Repellent plate, PS, Sterile, U Bottom, clear w/lid |