![greiner_bio_one955x590.jpg](https://occapi.avantorsciences.com/medias/greiner-bio-one955x590.jpg?context=bWFzdGVyfHJvb3R8ODQ3NDJ8aW1hZ2UvanBlZ3xhREk1TDJnMFpTODVORGM1TlRrd05EYzNPRFUwTDJkeVpXbHVaWEpmWW1sdlgyOXVaVGsxTlhnMU9UQXVhbkJufGM4MjgxZTg2MmFlZTU4MmNiMzRhMGY5NDk0ODhkMTc1NDkwNmY4MThlMGI2NTlkMDU2MzMwYzIzNmRlODQ0NWQ)
![greiner-bio-one-logo-60h.jpg](https://occapi.avantorsciences.com/medias/greiner-bio-one-logo-60h.jpg?context=bWFzdGVyfHJvb3R8MzA1OHxpbWFnZS9qcGVnfGFEWTJMMmcyTmk4NU5EYzVOVGMzTmpNeU56azRMMmR5WldsdVpYSXRZbWx2TFc5dVpTMXNiMmR2TFRZd2FDNXFjR2N8YmNiNGRhZGUwYjY1YWI3MGYzOWQ3ZDA1M2NiMTg5Zjk0YzY4MzVlY2YzOWI2OTBhMmI2YzFmMTYzZTEzODViZQ)
Greiner Bio-One
Greiner Bio-One, a world-renowned partner for academia, research establishments, diagnostic, pharmaceutical and biotechnology industries.
Integrating 50 years of injection-molding expertise with scientific innovation, all Greiner Bio-One products benefit from precision engineering, premium materials and advanced surface technologies.
A wide spectrum of flasks, dishes and multiwell microplates, with specialized surface chemistries and substrates that provide quality and consistency for individual cell and tissue culture needs.
![cellculture200x200.jpg](https://occapi.avantorsciences.com/medias/cellculture200x200.jpg?context=bWFzdGVyfGltYWdlc3w5MTEzfGltYWdlL2pwZWd8YURRNUwyZ3lNaTg1TlRBME5qQTBNRFl4TnpJMkwyTmxiR3hqZFd4MGRYSmxNakF3ZURJd01DNXFjR2N8Y2IyZGQwYmVhZjQ2NjZhYzRlZWQzNGFlMjQ0OTMzNmNjYTU1NTQ0YWU0ZDA4N2Q3YjAwYzkyMDQxYWE3OWViNg)
3D Cell Culture
The core technology is the magnetization of cells with NanoShuttle™-PL. The cells can be aggregated with magnetic forces, either by levitation or printing, to form structurally and biologically representative 3D models in vitro.
An extensive range of microplates for state of the art methods in Genomics, Proteomics, Cell Based, Biochemical and Immunoassay, specializing in HTS for Drug Discovery.
Greiner Bio-One manufactures 96-well ELISA and Strip Well Microplates, including Single Break, for diagnostic and immunological research applications within global life science industries.
Products designed tooptimize rapid, efficient density gradient centrifugation enrichment of Peripheral Blood Mononuclear Cells or Circulating Tumor Cells & filtration of cell or tissue suspensions.
A full line of premium quality liquid handling consumables including serologicals, pipette tips, and tubes that enable consistent and reliable research.