Refractometry & Polarimetry
Refractometry is one of the basic methods for quantitative analysis and quality assurance methods in food and beverage industry. Refractometers are frequently used by grape growers and the sugar industry for Brix testing of sucrose levels in their fruit.
For more precise results, polarimeters are used to measure optical rotation of the sucrose solutions. Avantor® offers a large selection of equipment and products for every refractometry and polarimetry application.
Product selection
The portable, easiest and most cost effective way to get a RI or Brix result...
For traditional measurements in refractometry...
Modern and precise refractometers - reliable, accurate, documentation with printouts and data storage..
Optical rotation at its best...
CertiPUR reference material for refractometry...