![mflx-LS-pump-series-1920x760.jpg](https://occapi.avantorsciences.com/medias/mflx-LS-pump-series-1920x760.jpg?context=bWFzdGVyfHJvb3R8OTUzNDR8aW1hZ2UvanBlZ3xhRGRsTDJneE5pODVORGM1TmpRMk5qYzBPVGMwTDIxbWJIZ3RURk10Y0hWdGNDMXpaWEpwWlhNdE1Ua3lNSGczTmpBdWFuQm58NzdkMDA3ZTcxMjFiMThhYzY1YmU5N2E2MjhiYjU2NDAxZTFmODkzMWU3MjQwYmZlYzM4MTI2ODFkYmQyNDk5ZQ)
Masterflex® L/S® Series Peristaltic Pumps
Versatility and precision
Accurate and reliable pumps featuring easy-to-use connectivity and control.
Flow range: 0.00005 to 3400 mL/min
Masterflex L/S Series peristaltic pumps include fixed and variable speeds, providing high accuracy drives for precise flow control and dispensing. Ideal for laboratory, process and field use.
Masterflex® L/S Series Pumps Features and Benefits
Our L/S series pumps are relied on for high accuracy and repeatable performance, plus advanced features expected in biopharmaceutical and other critical applications today, including: high-accuracy, precision-flowing and dispensing.
Our product features and benefits include:
- High volumetric efficiency
- Fluid only touches tubing
- Capability of operating dry with entrained air
- Multi-channel models for low pulsation requirement
- Intuitive control keypad
- Universal voltage
- Quick start guide for faster set-up
- Changing tubes takes 30 seconds
- Splash-resistant front panel
- Reversible pump direction flow
- Precision flow control
Our pumps are easy to use, easy to maintain, and ergonomically designed for ideal lab implementation.
Find over 20 pump tubing formulations across 10 L/S series compatible sizes. Precision engineered for the Masterflex® peristaltic pumps, giving the highest accuracy and longest life.
Reimage fluid handling
NEW integrated sensor technology, intuitive touchscreen and MasterflexLive® remote monitoring for the biopharma industry of tomorrow
Custom services and tools
Partnering on custom engineered pumps – making ideas reality for over 50 years.
We have the flexibility to meet your needs and save you time and money. Choose your tubing type, length and quantity.
With MasterflexLive®, you get real-time monitoring of all pump parameters.
Your source for disposable sterile bioprocess assemblies.
Precise, repeatable performance that is quick and easy to set up and program, IP66 stainless steel housing
A highly accurate IP66, stainless steel benchtop pump system for dosing, dispensing, and fill/finish applications
High precision, repeatable flow control, and reliable performance, you can have it all in one small footprint
Masterflex L/S Series Pump Components
Need more information or help selecting the perfect product for your application?
Our experts are ready to help. Contact us at 1-800-MASTERFLEX or [email protected]