BPy-PMO
Catalog # TCIAB5516-200MG
Supplier: TCI
CAS Number:
Specifications
- VPE:200 mg
- Seq:2530
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Specific surface:600 to 900 m2/g
- Pore volume:0.470 to 0.650 mL/g
- Pore diameter:4.00 to 4.50 nm
Specification Test Results
About this item
Synonyms: 2,2'-Bipyridyl linked Periodic Mesoporus Organosilica, Periodic Mesoporus Organosilica containing 2,2'-Bipyridine
: