Terpentijn olie, TECHNICAL
Catalogus # 23699.293
Leverancier: VWR Chemicals
CAS-nummer:
Specificaties
- Verpakking:Glass bottle
- VPE:1 L
- Seq:5037
- Environmentally Preferable:
Specificaties
Specificatie testresultaten
- Identification:Passes test
Specificatie testresultaten
Over dit artikel
Synoniemen: Oil of turpentine, Fir oil, Turpentine, Turpentine, Rectified, Pine oil, L-Turpentine
:
EINECS:232-350-7
UN:1299
BEREIKEN:01-2119553060-53
:GHS02
:GHS07
:GHS08
:GHS09
CAS-nummer:8006-64-2
MDL-nummer:MFCD00147790
Kokend Pt:153…175 °C (1013 hPa)
Smelten Pt:–60…–50 °C
Dichtheid:0.854…0.868 g/cm³ (25 °C)
Flits Pt:35
ADR:3 III