Specifications
- Type antilichaam:Cleavage And Polyadenylation Specific Factor 2, 100kDa
- antigeen symbool:CPSF2
- Clonality:Polyklonaal
- Conjugatie:Onconjugeerd
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Formulier:Lyophilized
- Antigeen synoniemen:cleavage and polyadenylation specificity factor subunit 2|CPSF100|cleavage and polyadenylation specificity factor 100 kDa subunit|CPSF 100 kDa subunit|CPSF 100kDa subunit
- Immunogeen:A synthetic peptide corresponding a region of human CPSF2.
- Maat:50 µg
- VPE:50 µG
Specifications
About this item
CPSF2 is a component of the cleavage and polyadenylation specificity factor (CPSF) complex that play a key role in pre-mRNA 3'-end formation, recognizing the AAUAAA signal sequence and interacting with poly (A) polymerase and other factors to bring about cleavage and poly (A) addition. It belongs to the metallo-beta-lactamase superfamily, RNA-metabolizing metallo-beta-lactamase-like family, CPSF2/YSH1 subfamily.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: CPSF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: