- Type antilichaam:Primair
- Clonality:Polyklonaal
- Conjugatie:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemie:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Kruisadsorptie:No
- Formulier:liquid
- Gen-ID:3017. 8339. 8343. 8344. 8346. 8347.
- Opslagbuffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Opslagtemperatuur:Store at 4°C for 12 months
- Concentratie:1 μg/μl
- Shipping temperature:4°C
- Immunogeen:1-50/126
- Purification:Purified by Protein A
- VPE:100 µl
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a member of the histone H2B family. Transcripts from this gene lack polyA tails; instead, they contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6p22-p21.3. [provided by RefSeq, Jul 2008].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Histone H2B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat