Human alpha-2-macroglobulin (from plasma)
: Biorbyt
- Bron:
- Species:
- Environmentally Preferable:
Frequently Bought Together
Nitril handschoenen, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Serologische pipetten, standaardreeks
: VWR Collection
Aceton ≥99%, TECHNICAL
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrilhandschoenen
: VWR Collection
VWR®, Transferpipetten, Hoge Prestaties
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Nitrilhandschoenen
: VWR Collection
VWR®, gezichtstissues
: VWR Collection
pH-teststrips, MQuant®, Supelco®
: Merck