Order Entry
Belgium
ContactUsLinkComponent
Anti-ASF Rabbit Polyclonal Antibody
Anti-ASF Rabbit Polyclonal Antibody
Catalogus # ORIGTA327123
Leverancier:  OriGene
Anti-ASF Rabbit Polyclonal Antibody
Catalogus # ORIGTA327123
Leverancier:  OriGene

Specificaties

  • Type antilichaam:
    Primair
  • antigeen symbool:
    ASF
  • Clonality:
    Polyklonaal
  • Conjugatie:
    Onconjugeerd
  • Host:
    Rabbit
  • ImmunoChemie:
    Yes
  • ImmunoFluorescentie:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Kruisadsorptie:
    No
  • Formulier:
    Liquid
  • Opslagbuffer:
    Store at -20C or -80C. Avoid freeze / thaw cycles. Buffer: PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
  • Opslagtemperatuur:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogeen:
    Recombinant protein of human SFRS1
  • Purification:
    purified by affinity chromatography
  • Maat:
    100 μg
  • VPE:
    100 µG

Specificaties

Over dit artikel

Recommended Dilutions: Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200; Immunofluorescence: 1:50-1:200

Type: Primary
Antigen: ASF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat