Anti-Calcium and Integrin Binding Protein 1 Mouse Monoclonal Antibody [clone: 6A14]
Catalog # USBIC0116-05B
Supplier: US Biological
Specifications
- Type antilichaam:Primair
- antigeen symbool:CIB1
- Clonality:Monoklonaal
- Clone:6A14
- Conjugatie:Onconjugeerd
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Formulier:liquid
- Opslagbuffer:PBS, pH 7,4, 0,1% sodium azide.
- Opslagtemperatuur:Cool
- Concentratie:0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Maat:100 µg
- VPE:100 µG
Specifications
About this item
Recommended dilution: ELISA: 0.1-1 ?g/ml; Western Blot: 1-3?g/ml
Type: Primary
Antigen: Calcium and Integrin Binding Protein 1
Clonality: Monoclonal
Clone: 6A14
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human