Anti-Superoxide Dismutase Copper Chaperone Mouse Monoclonal Antibody
Catalog # USBI136453
Supplier: US Biological
Specifications
- Type antilichaam:Primair
- Clonality:Monoklonaal
- Conjugatie:Onconjugeerd
- ELISA:Yes
- Host:Mouse
- ImmunoNeerslag:Yes
- Isotype:IgG2a kappa
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Formulier:liquid
- Opslagbuffer:HEPES with 0,15M NaCl, 0,01% BSA, 0,03% sodium azide, and 50% glycerol.
- Opslagtemperatuur:Cool
- Concentratie:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by ammonium sulfate precipitation
- Maat:50 µl
- VPE:50 µl
Specifications
About this item
Recommended dilution: :; Western Blot: 1:2000; Immunochemistry: 1:200
Type: Primary
Antigen: Superoxide Dismutase Copper Chaperone
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: