Anti-DENTT / CDA1 Goat polyclonal antibody unconjugated
Catalog # EVBIEB05240
Supplier: Everest Biotech
Specifications
- Type antilichaam:Primair
- antigeen symbool:DENTT
- Clonality:Polyklonaal
- Conjugatie:Onconjugeerd
- ELISA:Yes
- Host:Goat
- ImmunoChemie:Yes
- Isotype:IgG
- Reactivity:Human,Dog
- Environmentally Preferable:
- Epitoop:C-terminal
- Kruisadsorptie:none
- Antigeen synoniemen:CDA1
- Opslagtemperatuur:Cool
- Shipping temperature:Ice
- Maat:100 µg
- VPE:0,1 mg
Specifications
About this item
Type: Primary
Antigen: DENTT / CDA1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human