Anti-Heregulin/ZNDF/GGF/Neuregulin Mouse Monoclonal Antibody
Catalogus # BIRBORB108811-100
Leverancier: Biorbyt
Specificaties
- Type antilichaam:Primair
- antigeen symbool:ZNDF
- Clonality:Monoklonaal
- Conjugatie:Onconjugeerd
- Host:Mouse
- ImmunoChemie:Yes
- ImmunoNeerslag:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Antigeen synoniemen:Neuregulin|GGF
- Opslagtemperatuur:Cool
- Shipping temperature:Ice
- Immunogeen:Hybridoma produced by the fusion of spelocytes from BALB/c mice immunized with recombinant extracellular domain of rat NDF protein coupled to KLH and mouse myeloma NSO cells.
- Purification:purified by Protein A/G Chromatography
- Maat:100 µg
- VPE:100 µG
Specificaties
Over dit artikel
Type: Primary
Antigen: Heregulin/ZNDF/GGF/Neuregulin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat