Anti-CEBPZ Mouse Polyclonal Antibody
ABNOH00010153-B01P
: Abnova
- Type antilichaam:Primair
- antigeen symbool:CEBPZ
- Clonality:Polyklonaal
- Conjugatie:Onconjugeerd
- Host:Mouse
- ImmunoFluorescentie:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Kruisadsorptie:No
- Gen-ID:10153
- Antigeen synoniemen:NOC1|CCAAT box binding transcription factor|CBF2|CCAAT/enhancer binding protein zeta|CBF|HSPCBF|Cebpz|CCAAT binding factor
- Opslagtemperatuur:Cool
- Shipping temperature:Ice
- Maat:50 µg
- VPE:50 µg
Type: Primary
Antigen: CEBPZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat