Order Entry
Belgium
ContactUsLinkComponent
Anti-GDA Mouse Monoclonal Antibody [clone: 1D5]
Anti-GDA Mouse Monoclonal Antibody [clone: 1D5]
  ABNOH00009615-M01
 :  Abnova
undefined
Anti-GDA Mouse Monoclonal Antibody [clone: 1D5]
  ABNOH00009615-M01
 :  Abnova

 

  • Type antilichaam:
    Primair
  • antigeen symbool:
    GDA
  • Clonality:
    Monoklonaal
  • Clone:
    1D5
  • Conjugatie:
    Onconjugeerd
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Kruisadsorptie:
    No
  • Gen-ID:
    9615
  • Antigeen synoniemen:
    GUANASE|NEDASIN|CYPIN
  • Opslagtemperatuur:
    Cool
  • Shipping temperature:
    Ice
  • Maat:
    100 µg
  • VPE:
    100 µg

 

 

Type: Primary
Antigen: GDA
Clonality: Monoclonal
Clone: 1D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human