Anti-TMLHE Rabbit Polyclonal Antibody
Catalog # AVIVARP57145_P050
Supplier: Aviva Systems Biology
Specifications
- Type antilichaam:Primair
- antigeen symbool:TMLHE
- Clonality:Polyklonaal
- Conjugatie:Onconjugeerd
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitoop:middle
- Kruisadsorptie:none
- Gen-ID:55217
- Antigeen synoniemen:TMLHED|TMLD|TMLH|AUTSX6|BBOX2|XAP130
- Opslagtemperatuur:Cool
- Shipping temperature:Ice
- VPE:50 µG
Specifications
About this item
Type: Primary
Antigen: TMLHE (trimethyllysine hydroxylase, epsilon)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: