Order Entry
Belgium
ContactUsLinkComponent
Anti-Slow skeletal Myosin Rabbit Polyclonal Antibody
Anti-Slow skeletal Myosin Rabbit Polyclonal Antibody
  ANTIA101365-100
New Product
undefined
Anti-Slow skeletal Myosin Rabbit Polyclonal Antibody
  ANTIA101365-100
New Product

 

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    Cardiac myosin light chain 1
  • Symbole de l'antigène:
    Slow skeletal Myosin
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • ELISA:
    Yes
  • Hôte:
    Rabbit
  • Isotype:
    IgG
  • Réactivité:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Formulaire:
    Liquid
  • ID de gène:
    UniprotID# P08590
  • Synonymes antigène:
    Myosin light chain 3|MLC1V|CMLC1|MYL3_HUMAN|Myosin light chain 1 slow twitch muscle B ventricular isoform|myosin light chain 3 alkali ventricular skeletal slow|MYL3|MLC1SB|CMH8
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    21 kDa
  • Température de stockage:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Température de transport:
    Shipped on blue ice at 4 °C
  • Immunogène:
    Synthetic peptide derived from human MYL3 (amino acids 71 - 120)
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
  • Type de conditionnement:
    Plastic vial
  • Cdt:
    100 µG

 

 

Rabbit polyclonal antibody to MYL3 for WB and ELISA with samples derived from human.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000

Type: Primary
Antigen: Slow skeletal Myosin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human