Anti-Nephrocystin 4 Rabbit Polyclonal Antibody
N° de catalogue ANTIA100655-100
Fournisseur: ANTIBODIES.COM
New Product
Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:KIAA0673
- Symbole de l'antigène:Nephrocystin 4
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:IgG
- Réactivité:Human
- Environmentally Preferable:
- Formulaire:Liquid
- ID de gène:UniprotID# O75161
- Synonymes antigène:POC10|Nephrocystin-4|SLSN4|POC10 centriolar protein homolog|nephronophthisis 4|Nephroretinin|Nephrocystin 4|NPHP4_HUMAN|NPHP4
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:157 kDa
- Température de stockage:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Température de transport:Shipped on blue ice at 4 °C
- Immunogène:Synthetic peptide derived from human NPHP4 (amino acids 877 - 926)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Type de conditionnement:Plastic vial
- Cdt:100 µG
Spécifications
À propos de cet article
Rabbit polyclonal antibody to NPHP4 for IHC and ELISA with samples derived from human.
- Validated Applications: IHC, ELISA
- Recommended Dilutions: IHC: 1:50 to 1:100, ELISA: 1:40000
Type: Primary
Antigen: Nephrocystin 4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human