Anti-SLC27A4/FATP4 Rabbit Polyclonal Antibody
Catalog # ANTIA100104-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:ACSVL 4
- Symbole de l'antigène:SLC27A4/FATP4
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Réactivité:Human
- Environmentally Preferable:
- Formulaire:Liquid
- ID de gène:UniprotID# Q6P1M0
- Synonymes antigène:EC 6.2.1|Fatty acid transport protein4|Long chain fatty acid transport protein4|Long chain fatty acid transport protein 4|ACSVL4|IPS|Fatty acid transport protein 4|FATP4|FATP 4
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:72 kDa
- Température de stockage:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Température de transport:Shipped on blue ice at 4 °C
- Immunogène:Synthetic peptide derived from human SLC27A4 (amino acids 61 - 110)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Type de conditionnement:Plastic vial
- Cdt:100 µG
Specifications
About this item
Rabbit polyclonal antibody to SLC27A4 for IF and ELISA with samples derived from human.
- Validated Applications: IF, ELISA
- Recommended Dilutions: IF: 1:100 to 1:500, ELISA: 1:20000
Type: Primary
Antigen: SLC27A4 / FATP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human