Order Entry
Belgium
ContactUsLinkComponent
Anti-KiSS1 receptor Rabbit Polyclonal Antibody
Anti-KiSS1 receptor Rabbit Polyclonal Antibody
N° de catalogue ANTIA100516-100
Fournisseur:  ANTIBODIES.COM
New Product
undefined
Anti-KiSS1 receptor Rabbit Polyclonal Antibody
N° de catalogue ANTIA100516-100
Fournisseur:  ANTIBODIES.COM
New Product

Spécifications

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    AXOR 12
  • Symbole de l'antigène:
    KiSS1 receptor
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • ELISA:
    Yes
  • Hôte:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Réactivité:
    Human
  • Environmentally Preferable:
  • Formulaire:
    Liquid
  • ID de gène:
    UniprotID# Q969F8
  • Synonymes antigène:
    AXOR12|GPCR54|GPCR 54|GPR 54|GPR54|G-protein coupled receptor 54|hOT7T175|G protein coupled receptor 54|G-protein coupled receptor OT7T175
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    42 kDa
  • Température de stockage:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Température de transport:
    Shipped on blue ice at 4 °C
  • Immunogène:
    Synthetic peptide derived from human KISS1R (amino acids 301-350)
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
  • Type de conditionnement:
    Plastic vial
  • Cdt:
    100 µG

Spécifications

À propos de cet article

Rabbit polyclonal antibody to KISS1R for IF and ELISA with samples derived from human.

  • Validated Applications: IF, ELISA
  • Recommended Dilutions: IF: 1:100 to 1:500, ELISA: 1:5000

Type: Primary
Antigen: KiSS1 receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human