Order Entry
Belgium
ContactUsLinkComponent
Anti-SLC39A1 Rabbit Polyclonal Antibody
Anti-SLC39A1 Rabbit Polyclonal Antibody
Catalog # ANTIA100097-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-SLC39A1 Rabbit Polyclonal Antibody
Catalog # ANTIA100097-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    hZIP1
  • Symbole de l'antigène:
    SLC39A1
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • ELISA:
    Yes
  • Hôte:
    Rabbit
  • Isotype:
    IgG
  • Réactivité:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Formulaire:
    Liquid
  • ID de gène:
    UniprotID# Q9NY26
  • Synonymes antigène:
    ZIP-1|IRT1|SLC39A1|Zinc transporter ZIP1|Zinc-iron-regulated transporter-like|ZIRTL|Zrt- and Irt-like protein 1|S39A1_HUMAN|Solute carrier family 39 member 1
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    34 kDa
  • Température de stockage:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Température de transport:
    Shipped on blue ice at 4 °C
  • Immunogène:
    Synthetic peptide derived from human SLC39A1 (amino acids 111-160)
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
  • Type de conditionnement:
    Plastic vial
  • Cdt:
    100 µG

Specifications

About this item

Rabbit polyclonal antibody to SLC39A1 for WB and ELISA with samples derived from human.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:5000

Type: Primary
Antigen: SLC39A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human