Anti-MEK3 + MEK6 Rabbit Polyclonal Antibody
ANTIA94431-100
New Product
- Type d'anticorps:Primaire
- Nom de l'antigène:Dual specificity mitogen activated protein kinase kinase 6
- Symbole de l'antigène:MEK3 + MEK6
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Liquid
- ID de gène:UniprotID# P46734
- Synonymes antigène:MAP kinase kinase 3|MAP kinase kinase 6|map2k3|MAP2K6|MAPK/ERK kinase 6|MAPKK 3|MAPK/ERK kinase 3|MAPKK 6|Dual specificity mitogen-activated protein kinase kinase 3
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:39 kDa
- Température de stockage:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Température de transport:Shipped on blue ice at 4 °C
- Immunogène:Synthetic peptide derived from human MKK3 around the phosphorylation site of Ser189 (amino acids 173-222)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Type de conditionnement:Plastic vial
- Cdt:100 µG
Rabbit polyclonal antibody to MKK3 (phospho Ser189) for WB, IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: WB, IHC, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:5000
Type: Primary
Antigen: MEK3 + MEK6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat