Order Entry
Belgium
ContactUsLinkComponent
Anti-MAPKAPK2/3 Rabbit Polyclonal Antibody
Anti-MAPKAPK2/3 Rabbit Polyclonal Antibody
N° de catalogue ANTIA93464-100
Fournisseur:  ANTIBODIES.COM
New Product
undefined
Anti-MAPKAPK2/3 Rabbit Polyclonal Antibody
N° de catalogue ANTIA93464-100
Fournisseur:  ANTIBODIES.COM
New Product

Spécifications

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    3PK
  • Symbole de l'antigène:
    MAPKAPK2/3
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • ELISA:
    Yes
  • Hôte:
    Rabbit
  • ImmunoChimie:
    Yes
  • Isotype:
    IgG
  • Réactivité:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Formulaire:
    Liquid
  • ID de gène:
    UniprotID# P49137
  • Synonymes antigène:
    MK2|AA960234|MAPK activated protein kinase 2|MAPKAP kinase 2|MAP kinase activated protein kinase 3|MAP kinase activated protein kinase 2|MAPKAP kinase 3|MAPKAPK2/3|MAPK activated protein kinase 3
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    45 kDa
  • Température de stockage:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Température de transport:
    Shipped on blue ice at 4 °C
  • Immunogène:
    Synthetic peptide derived from human MAPKAPK-2 around the phosphorylation site of Thr222 (amino acids 188-237)
  • Tested applications:
    IHC
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
  • Type de conditionnement:
    Plastic vial
  • Cdt:
    100 µG

Spécifications

À propos de cet article

Rabbit polyclonal antibody to MAPKAPK-2 (phospho Thr222) for WB, IHC and ELISA with samples derived from Human, Mouse and Rat.

  • Validated applications: WB, IHC, ELISA
  • Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:20000

Type: Primary
Antigen: MAPKAPK2/3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat