Order Entry
Belgium
ContactUsLinkComponent
 
Anti-Pdha2 Rabbit Polyclonal Antibody
  ANTIA93155-100
New Product
undefined
Anti-Pdha2 Rabbit Polyclonal Antibody
  ANTIA93155-100
New Product

 

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    EC 1.2.4.1
  • Symbole de l'antigène:
    Pdha2
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • Hôte:
    Rabbit
  • ImmunoChimie:
    Yes
  • Isotype:
    IgG
  • Réactivité:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Formulaire:
    Liquid
  • ID de gène:
    UniprotID# P35487
  • Synonymes antigène:
    PDHAL|PDHE1 A type II|Pyruvate dehydrogenase (lipoamide) alpha 2|MGC149517|PDHA2|ODPAT_HUMAN|MGC149518|mitochondrial|PDHE1-A type II
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,01% Thiomersal
  • Molecular weight:
    43 kDa
  • Sequence:
    ISYRSREEVHNVRSKSDPIMLLRERIISNNLSNIEELKEIDADVKKEVEDAAQFATTDPEPAVEDIANYLYHQDPPFEVRGAHKWLKYKSHS
  • Température de stockage:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Température de transport:
    Shipped on blue ice at 4 °C
  • Immunogène:
    A synthetic peptide corresponding to a sequence within amino acids 300 to the C-terminus of mouse PDHA2 (NP_032837.1)
  • Tested applications:
    IHC
  • Purification:
    Affinity purification
  • Type de conditionnement:
    Plastic vial
  • Cdt:
    100 µl

 

 

Rabbit polyclonal antibody to Pdha2 for WB and IHC with samples derived from Mouse and Rat.

  • Validated applications: WB, IHC
  • Recommended dilutions: WB: 1:500 to 1:2,000, IHC: 1:50 to 1:200

Type: Primary
Antigen: Pdha2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat