Order Entry
Belgium
ContactUsLinkComponent
 
Anti-LY75 Mouse Monoclonal Antibody [Clone: CD205/3720]
Catalog # ANTIA278285-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-LY75 Mouse Monoclonal Antibody [Clone: CD205/3720]
Catalog # ANTIA278285-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    C-type lectin domain family 13 member B
  • Symbole de l'antigène:
    LY75
  • Clonalité:
    Monoclonal
  • Clone:
    CD205/3720
  • Conjugaison:
    Unconjugated
  • Hôte:
    Mouse
  • ImmunoChimie:
    Yes
  • Isotype:
    IgG2b kappa
  • Réactivité:
    Human
  • Environmentally Preferable:
  • Formulaire:
    Liquid
  • ID de gène:
    UniprotID# O60449
  • Synonymes antigène:
    gp200|CLEC 13B|CD205|CD 205|DEC205|DEC 205|DEC-205|CD205 antigen|CLEC13B
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
  • Molecular weight:
    198 kDa
  • Température de stockage:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Température de transport:
    Shipped on blue ice at 4 °C
  • Immunogène:
    Recombinant fragment, around amino acids 1,006-1,130, of human LY75 protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Type de conditionnement:
    Plastic vial
  • Cdt:
    100 µG

Specifications

About this item

Mouse monoclonal [CD205/3720] antibody to LY75 for IHC-P with samples derived from human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: LY75
Clonality: Monoclonal
Clone: CD205/3720
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human