Anti-Cadherin 8 Rabbit Polyclonal Antibody
Catalog # ANTIA96228-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:CADH8_HUMAN
- Symbole de l'antigène:Cadherin 8
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Liquid
- ID de gène:UniprotID# P55286
- Synonymes antigène:Cadherin 8|Cdh8|Nbla04261|Cadherin 8, type 2|Putative protein product of Nbla04261|Cadherin-8|CDH 8
- Storage buffer:Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
- Molecular weight:88 kDa
- Température de stockage:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Température de transport:Shipped on blue ice at +4 °C
- Immunogène:Synthetic peptide derived from human CDH8 (amino acids 470-519)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Type de conditionnement:Plastic vial
- Cdt:100 µG
Specifications
About this item
Rabbit polyclonal antibody to CDH8 for WB, IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: WB, IHC, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:10000
Type: Primary
Antigen: Cadherin 8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat