Anti-ER1 Rabbit Polyclonal Antibody
ANTIA309740-100
New Product
- Type d'anticorps:Primaire
- Nom de l'antigène:ER1
- Symbole de l'antigène:ER1
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Oryza sativa
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Liquid
- ID de gène:UniprotID# Q69SP5
- Synonymes antigène:Mesoderm induction early response 1|Mesoderm induction early response 1 homolog (Xenopus laevis)|MI-ER1|MIER1
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol and 0,01% thiomersal
- Molecular weight:107 kDa
- Température de stockage:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Température de transport:Shipped on blue ice at 4 °C
- Immunogène:Recombinant protein of Oryza sativa OsERECTA.
- Purification:Affinity purification
- Type de conditionnement:Plastic vial
- Cdt:100 µl
Rabbit polyclonal antibody to ER1 for WB with samples derived from Oryza sativa.
- Validated applications: WB
- Recommended dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: ER1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Oryza sativa