Anti-MIRO1 Rabbit Polyclonal Antibody
Catalog # ANTIA309670-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:ARHT 1
- Symbole de l'antigène:MIRO1
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Liquid
- ID de gène:UniprotID# Q8IXI2
- Synonymes antigène:hMiro1|hMiro-1|Mitochondrial Rho 1|hMiro 1|MIRO1_HUMAN|ARHT1|mitochondrial Rho (MIRO) GTPase 1|FLJ12633|FLJ11040
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol and 0,05% Proclin 300
- Molecular weight:75 kDa
- Température de stockage:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Température de transport:Shipped on blue ice at 4 °C
- Immunogène:Recombinant protein of human RHOT1/MIRO1.
- Tested applications:ICC/IF
- Purification:Affinity purification
- Type de conditionnement:Plastic vial
- Cdt:100 µl
Specifications
About this item
Rabbit polyclonal antibody to MIRO1 for WB and ICC/IF with samples derived from human, mouse and rat.
- Validated applications: WB, ICC/IF
- Recommended dilutions: WB: 1:100 to 1:500, ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: MIRO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat