Anti-LKB1 Rabbit Polyclonal Antibody
ANTIA98064-100
New Product
- Type d'anticorps:Primaire
- Nom de l'antigène:hLKB1
- Symbole de l'antigène:LKB1
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:IgG
- Réactivité:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Liquid
- ID de gène:UniprotID# Q15831
- Synonymes antigène:Serine/Threonine Kinase 11|Serine/threonine protein kinase LKB1|Polarization related protein LKB1|PJS|Serine/threonine protein kinase 11|Renal carcinoma antigen NY-REN-19|Serine/threonine-protein kinase 11|Serine/threonine protein kinase STK11|Liver kinase B1
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:48 kDa
- Température de stockage:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Température de transport:Shipped on blue ice at +4 °C
- Immunogène:Synthetic peptide derived from human LKB1 (amino acids 384-433)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Type de conditionnement:Plastic vial
- Cdt:100 µG
Rabbit polyclonal antibody to LKB1 for WB, IHC and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, IHC, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:10000
Type: Primary
Antigen: LKB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse