Order Entry
Belgium
ContactUsLinkComponent
Anti-UBFD1 Rabbit Polyclonal Antibody
Anti-UBFD1 Rabbit Polyclonal Antibody
Catalog # ANTIA97126-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-UBFD1 Rabbit Polyclonal Antibody
Catalog # ANTIA97126-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    UBFD1
  • Symbole de l'antigène:
    UBFD1
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • ELISA:
    Yes
  • Hôte:
    Rabbit
  • Isotype:
    IgG
  • Réactivité:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Formulaire:
    Liquid
  • ID de gène:
    UniprotID# O14562
  • Synonymes antigène:
    Ubiquitin domain-containing protein UBFD1|UBFD1_HUMAN|UBPH|Ubiquitin-binding protein homolog
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    56 kDa
  • Température de stockage:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Température de transport:
    Shipped on blue ice at +4 °C
  • Immunogène:
    Synthetic peptide derived from human UBFD1 (amino acids 171-220)
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
  • Type de conditionnement:
    Plastic vial
  • Cdt:
    100 µG

Specifications

About this item

Rabbit polyclonal antibody to UBFD1 for WB and ELISA with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000

Type: Primary
Antigen: UBFD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat