Anti-Adenylate Cyclase 3 Rabbit Polyclonal Antibody
Fournisseur: ANTIBODIES.COM
New Product
Certificats
À propos de cet article
Rabbit polyclonal antibody to Adenylate Cyclase 3 for WB, ICC/IF and IHC with samples derived from human, rat and mouse.
- Validated applications: WB, ICC/IF, IHC
- Recommended dilutions: WB: 1:1000, ICC/IF: 1:10000 to 1:20000, IHC: 1:10000 to 1:2000
Type: Primary
Antigen: Adenylate Cyclase 3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat;Mouse
Spécifications
- Catalog No:
- ANTIA85287-50
- ANTIA85287-100
- Symbole de l'antigène:
- Adenylate Cyclase 3
- Adenylate Cyclase 3
- Nom de l'antigène:
- AC-III
- AC-III
- Synonymes antigène:
- ADCY3|Adenylate cyclase type III|ATP pyrophosphate lyase|ADCY3_HUMAN|AC3|adenylate cyclase 3|Adenylyl cyclase 3|Adenylate cyclase type 3|Adenylate cyclase
- ADCY3|Adenylate cyclase type III|ATP pyrophosphate lyase|ADCY3_HUMAN|AC3|adenylate cyclase 3|Adenylyl cyclase 3|Adenylate cyclase type 3|Adenylate cyclase
- Type d'anticorps:
- Primaire
- Primaire
- Clonalité:
- Polyclonal
- Polyclonal
- Conjugaison:
- Unconjugated
- Unconjugated
- Réactivité:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Hôte:
- Rabbit
- Rabbit
- ID de gène:
- UniprotID# P21932
- UniprotID# P21932
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChimie:
- Yes
- Yes
- Immunogène:
- Recombinant construct corresponding to the C-terminal, amino acids 1125-1144, of rat Adenylate Cyclase 3
- Recombinant construct corresponding to the C-terminal, amino acids 1125-1144, of rat Adenylate Cyclase 3
- Sequence:
- PAAFPNGSSVTLPHQVVDNP
- PAAFPNGSSVTLPHQVVDNP
- Formulaire:
- Liquid
- Liquid
- Molecular weight:
- 130-200 kDa (by SDS-PAGE)
- 130-200 kDa (by SDS-PAGE)
- Purification:
- Immunogen affinity purification.
- Immunogen affinity purification.
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage buffer:
- Supplied in phosphate buffered saline with 50% Glycerol and 5mM sodium azide
- Supplied in phosphate buffered saline with 50% Glycerol and 5mM sodium azide
- Température de stockage:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Température de transport:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C
- Tested applications:
- ICC/IF, IHC
- ICC/IF, IHC
Spécifications
Frequently Bought Together