Order Entry
Belgium
ContactUsLinkComponent
 
Anti-NDUFA7 Goat Polyclonal Antibody
  ANTIA85062-100
New Product
undefined
Anti-NDUFA7 Goat Polyclonal Antibody
  ANTIA85062-100
New Product

 

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    B14.5a
  • Symbole de l'antigène:
    NDUFA7
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • ELISA:
    Yes
  • Hôte:
    Goat
  • ImmunoChimie:
    Yes
  • Isotype:
    IgG
  • Réactivité:
    Pig,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Formulaire:
    Liquid
  • Synonymes antigène:
    NADH ubiquinone oxidoreductase subunit B14.5a|NDUFA7|NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 7 14.5kDa|CI-B14.5a|Complex I B14.5a|NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 7|NADH-ubiquinone oxidoreductase subunit B14.5a|NDUA7_HUMAN|Complex I-B14.5a
  • Storage buffer:
    Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
  • Sequence:
    C-PKLPVGPSHKLSNN
  • Température de stockage:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    100 µg at 0,5 mg/ml
  • Température de transport:
    Shipped on blue ice at 4 °C
  • Immunogène:
    Synthetic peptide corresponding to Human NDUFA7 (internal region)
  • Tested applications:
    IHC
  • Purification:
    Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
  • Type de conditionnement:
    Plastic vial
  • Cdt:
    100 µG

 

 

Goat polyclonal antibody to NDUFA7 for ELISA, WB and IHC with samples derived from human, mouse, rat and porcine.

  • Validated applications: ELISA, WB, IH

Type: Primary
Antigen: NDUFA7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Mouse;Rat;Porcine