Order Entry
Belgium
ContactUsLinkComponent
 
Anti-Foxi3 Goat Polyclonal Antibody
  ANTIA84944-100
New Product
undefined
Anti-Foxi3 Goat Polyclonal Antibody
  ANTIA84944-100
New Product

 

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    Forkhead box I3
  • Symbole de l'antigène:
    Foxi3
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • ELISA:
    Yes
  • Hôte:
    Goat
  • Isotype:
    IgG
  • Réactivité:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Formulaire:
    Liquid
  • Synonymes antigène:
    FOXI3|LOC344167|FOXI3_HUMAN|Forkhead box protein I3
  • Storage buffer:
    Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
  • Sequence:
    C-RLRVSGKLEGDSPS
  • Température de stockage:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    100 µg at 0,5 mg/ml
  • Température de transport:
    Shipped on blue ice at 4 °C
  • Immunogène:
    Synthetic peptide corresponding to Mouse Foxi3 (internal region)
  • Purification:
    Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
  • Type de conditionnement:
    Plastic vial
  • Cdt:
    100 µG

 

 

Goat polyclonal antibody to FOXI3 for ELISA and WB with samples derived from mouse and rat.

  • Validated applications: ELISA, W

Type: Primary
Antigen: Foxi3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse;Rat