Anti-VTI1B Goat Polyclonal Antibody
Catalog # ANTIA84202-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:HGNC:17793
- Symbole de l'antigène:VTI1B
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Goat
- Isotype:IgG
- Réactivité:Human
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Liquid
- Synonymes antigène:v-SNARE|VTI1 like|VTI1|Vesicle associated soluble NSF attachment protein receptor|V SNARE|Vesicle transport through interaction with t SNAREs 1B|Vesicle transport through interaction with t-SNAREs homolog 1B|Vesicle transport v-SNARE protein Vti1-like 1|Vesicle transport through interaction with t SNAREs homolog 1B (yeast)
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
- Sequence:C-RDFDEKQQEANET
- Température de stockage:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Température de transport:Shipped on blue ice at 4 °C
- Immunogène:Synthetic peptide corresponding to Human VTI1B (internal region)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Type de conditionnement:Plastic vial
- Cdt:100 µG
Specifications
About this item
Goat polyclonal antibody to VTI1B for ELISA and WB with samples derived from human.
- Validated applications: ELISA, W
Type: Primary
Antigen: VTI1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human