Anti-RIC8A Goat Polyclonal Antibody
ANTIA84046-100
New Product
- Type d'anticorps:Primaire
- Nom de l'antigène:MGC104517
- Symbole de l'antigène:RIC8A
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Goat
- Isotype:IgG
- Réactivité:Human
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Liquid
- Synonymes antigène:Synembryn-A|RIC8|RIC8A_HUMAN|Protein Ric-8A|MGC148073|MGC148074|MGC131931|RIC8A|resistance to inhibitors of cholinesterase 8 homolog A (C. elegans)
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
- Sequence:CLSSDPDSDPD
- Température de stockage:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Température de transport:Shipped on blue ice at 4 °C
- Immunogène:Synthetic peptide corresponding to Human RIC8A (C terminal)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Type de conditionnement:Plastic vial
- Cdt:100 µG
Goat polyclonal antibody to RIC8A for ELISA and WB with samples derived from human.
- Validated applications: ELISA, W
Type: Primary
Antigen: RIC8A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human