Anti-pro Caspase1 + p10 + p12 Rabbit Polyclonal Antibody
ANTIA99669-100
New Product
- Type d'anticorps:Primaire
- Nom de l'antigène:CASP1
- Symbole de l'antigène:pro Caspase1 + p10 + p12
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Liquid
- ID de gène:UniprotID# P29466
- Synonymes antigène:IL1B convertase|pro Caspase1 + p10 + p12|ICE|IL 1BC|Caspase 1 apoptosis related cysteine peptidase|IL 1 beta converting enzyme|Interleukin 1 beta converting enzyme|p45|Interleukin 1 beta convertase
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:35 kDa
- Température de stockage:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Température de transport:Shipped on blue ice at 4 °C
- Immunogène:Synthetic peptide derived from the C-terminal region of human CASP1 (amino acids 350-400)
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific peptide
- Type de conditionnement:Plastic vial
- Cdt:100 µG
Rabbit polyclonal antibody to CASP1 for WB and ELISA with samples derived from human and rat.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: pro Caspase1 + p10 + p12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat