Order Entry
Belgium
ContactUsLinkComponent
Anti-DAP1 Rabbit Polyclonal Antibody
Anti-DAP1 Rabbit Polyclonal Antibody
N° de catalogue ANTIA99481-100
Fournisseur:  ANTIBODIES.COM
New Product
undefined
Anti-DAP1 Rabbit Polyclonal Antibody
N° de catalogue ANTIA99481-100
Fournisseur:  ANTIBODIES.COM
New Product

Spécifications

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    Dap
  • Symbole de l'antigène:
    DAP1
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • ELISA:
    Yes
  • Hôte:
    Rabbit
  • ImmunoChimie:
    Yes
  • Isotype:
    IgG
  • Réactivité:
    Human,
    Mouse
  • Environmentally Preferable:
  • Formulaire:
    Liquid
  • ID de gène:
    UniprotID# P51397
  • Synonymes antigène:
    DAP1_HUMAN|DAP1|Death associated protein 1|DAP 1|DAP-1|MGC99796|Death associated protein|OTTHUMP00000161670|Death-associated protein 1
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    11 kDa
  • Température de stockage:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Température de transport:
    Shipped on blue ice at 4 °C
  • Immunogène:
    Synthetic peptide derived from human DAP (amino acids 11-60)
  • Tested applications:
    IHC
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
  • Type de conditionnement:
    Plastic vial
  • Cdt:
    100 µG

Spécifications

À propos de cet article

Rabbit polyclonal antibody to DAP for IHC and ELISA with samples derived from human and mouse.

  • Validated applications: IHC, ELISA
  • Recommended dilutions: IHC: 1:50 to 1:100, ELISA: 1:40000

Type: Primary
Antigen: DAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse