Anti-ITCH/AIP4 Rabbit Polyclonal Antibody
ANTIA99399-100
New Product
- Type d'anticorps:Primaire
- Nom de l'antigène:ADMFD
- Symbole de l'antigène:ITCH/AIP4
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Liquid
- ID de gène:UniprotID# Q96J02
- Synonymes antigène:AIF4|dJ468O1.1 (atrophin 1 interacting protein 4 (AIP4))|AIP4|Atrophin 1 interacting protein 4|E3 ubiquitin protein ligase Itchy homolog|E3 ubiquitin-protein ligase Itchy homolog|dJ468O1.1|Atrophin-1-interacting protein 4|dJ468O1.1 atrophin 1 interacting protein 4 AIP4
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:102 kDa
- Température de stockage:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Température de transport:Shipped on blue ice at 4 °C
- Immunogène:Synthetic peptide derived from human ITCH (amino acids 386-435)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Type de conditionnement:Plastic vial
- Cdt:100 µG
Rabbit polyclonal antibody to ITCH for WB and ELISA with samples derived from human and mouse.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:40000
Type: Primary
Antigen: ITCH/AIP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse