Anti-gamma Adducin Rabbit Polyclonal Antibody
ANTIA98300-100
New Product
- Type d'anticorps:Primaire
- Nom de l'antigène:ADD3
- Symbole de l'antigène:gamma Adducin
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Liquid
- ID de gène:UniprotID# Q9UEY8
- Synonymes antigène:Adducin 3|ADDG_HUMAN|Gamma-adducin|ADDL|Adducin 3 (gamma)|Adducin-like|gamma Adducin|Adducin-like protein 70|Adducin, gamma
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:79 kDa
- Température de stockage:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Température de transport:Shipped on blue ice at 4 °C
- Immunogène:Synthetic peptide derived from human ADD3 (amino acids 403-452)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Type de conditionnement:Plastic vial
- Cdt:100 µG
Rabbit polyclonal antibody to ADD3 for WB and ELISA with samples derived from human, mouse and rat.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: gamma Adducin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat